missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-7 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-380AF750
This item is not returnable.
View return policy
Description
Galectin-7 Polyclonal antibody specifically detects Galectin-7 in Mouse samples. It is validated for Western Blot
Specifications
| Galectin-7 | |
| Polyclonal | |
| Western Blot | |
| Gal-7, GAL7, galectin 7, galectin-7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
| The immunogen recognized by this antibody maps to a region between residue 100 and the C-terminus (residue 135) of human Galectin-7 using the numbering given in entry NP_002298.1 (GeneID 3963). | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 3963 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction