missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-9 Antibody (OTI1D12), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71132AF350
This item is not returnable.
View return policy
Description
Galectin-9 Monoclonal specifically detects Galectin-9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Galectin-9 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LGALS9 | |
| Full length recombinant protein of human Galectin-9 (NP_002299) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1D12 | |
| Western Blot, Immunohistochemistry | |
| ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
| Mouse | |
| Affinity Purified | |
| Myeloid derived Suppressor Cell, Signal Transduction | |
| 3965.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction