missing translation for 'onlineSavingsMsg'
Learn More

Galectin-9 Antibody (RG9-35) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody

€ 262.00 - € 641.00

Specifications

Antigen Galectin-9
Clone RG9-35
Concentration 1 mg/mL
Applications Flow Cytometry, Immunofluorescence, Neutralization, Functional Assay
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18693707
View Documents
Novus Biologicals
NBP3-09032-0.025mg
0.025 mg
€ 262.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18613519
View Documents
Novus Biologicals
NBP3-09032-0.2mg
0.2 mg
€ 641.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Neutralization, Functional Assay
Specifications

Specifications

Galectin-9
1 mg/mL
Monoclonal
Purified
RUO
Mouse
ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
This antibody was raised by immunising rats with recombinant mouse galectin-9.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
RG9-35
Flow Cytometry, Immunofluorescence, Neutralization, Functional Assay
Unconjugated
Rabbit
Myeloid derived Suppressor Cell, Signal Transduction
PBS
3965
IgG κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.