missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561469
This item is not returnable.
View return policy
Description
Immunogen sequence: RFDENAVVRN TQIDNSWGSE ERSLPRKMPF VRGQSFSVWI LCEAHCLKVA VDGQHLFEYY HRLRNLPTIN Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin's disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
| Galectin 9 | |
| Polyclonal | |
| Unconjugated | |
| Lgals9 | |
| 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; tumor antigen HOM-HD-21; UAT; urate transporter/channel; urate transporter/channel protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3965 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00182 | |
| Lgals9 | |
| Recombinant protein corresponding to Human Galectin 9. Recombinant protein control fragment (Product #RP-99534). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction