missing translation for 'onlineSavingsMsg'
Learn More

Gallium, plasma standard solution, Specpure™ Ga 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4824717
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-55-3, 7647-01-0, 7732-18-5
69.72
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
5360835
CHEBI:49631
gallium
[Ga]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-55-3, 7647-01-0, 7732-18-5
69.72
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
5360835
CHEBI:49631
gallium
[Ga]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Molecular Formula Ga
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Gallium: 0.1%; Hydrochloric acid: <0.1%; Water: 94.8%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Gallium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'