missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALM Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97206AF594
This item is not returnable.
View return policy
Description
GALM Polyclonal antibody specifically detects GALM in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GALM | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| aldose 1-epimerase, BLOCK 25, EC 5.1.3.3, galactomutarotase, Galactose mutarotase, galactose mutarotase (aldose 1-epimerase), IBD1 | |
| Produced in rabbits immunized with E. coli-derived Human GALM fragment. | |
| 0.1 mL | |
| Cell Biology, Lipid and Metabolism | |
| 130589 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction