missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALM Monoclonal Antibody (OTI7B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812291S
This item is not returnable.
View return policy
Description
GALM Monoclonal antibody specifically detects GALM in Human samples. It is validated for Western BlotSpecifications
| GALM | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BW66 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51550 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7B7 | |
| Unconjugated | |
| CINP | |
| A530057M15Rik; aldose 1-epimerase; aldose mutarotase; AU015645; AU020959; BLOCK25; epididymis secretory sperm binding protein Li 63p; galactomutarotase; galactose enzyme activator; galactose mutarotase; galactose mutarotase (aldose 1-epimerase); GALM; GLAT; HEL-S-63p; IBD1 | |
| Full length human recombinant protein of human CINP produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction