missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALM Monoclonal Antibody (OTI7B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812291
This item is not returnable.
View return policy
Description
This gene encodes an enzyme that catalyzes the epimerization of hexose sugars such as glucose and galactose. The encoded protein is expressed in the cytoplasm and has a preference for galactose. The encoded protein may be required for normal galactose metabolism by maintaining the equilibrium of alpha and beta anomers of galactose.Specifications
| GALM | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96C23 | |
| GALM | |
| Full length human recombit protein of human GALM produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7B7 | |
| Unconjugated | |
| GALM | |
| BLOCK25, GLAT, HEL-S-63p, IBD1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 130589 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction