missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557284
This item is not returnable.
View return policy
Description
Immunogen sequence: YPGELKVWVT YTLDGGELIV NYRAQASQAT PVNLTNHSYF NLAGQASPNI NDHEVTIEAD TYLPVDETLI PTGEVAPVQG TAFDLRK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
This gene encodes an enzyme that catalyzes the epimerization of hexose sugars such as glucose and galactose. The encoded protein is expressed in the cytoplasm and has a preference for galactose. The encoded protein may be required for normal galactose metabolism by maintaining the equilibrium of alpha and beta anomers of galactose.
Specifications
| GALM | |
| Polyclonal | |
| Unconjugated | |
| GALM | |
| A530057M15Rik; aldose 1-epimerase; aldose mutarotase; AU015645; AU020959; BLOCK25; epididymis secretory sperm binding protein Li 63p; galactomutarotase; galactose enzyme activator; galactose mutarotase; galactose mutarotase (aldose 1-epimerase); GALM; GLAT; HEL-S-63p; IBD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 130589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96C23 | |
| GALM | |
| Recombinant protein corresponding to Human GALM. Recombinant protein control fragment (Product #RP-96110). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction