missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GALM Polyclonal Antibody
GREENER_CHOICE

Product Code. 16620045 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16620045 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16620045 Supplier Invitrogen™ Supplier No. PA5103591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GALM.

This gene encodes an enzyme that catalyzes the epimerization of hexose sugars such as glucose and galactose. The encoded protein is expressed in the cytoplasm and has a preference for galactose. The encoded protein may be required for normal galactose metabolism by maintaining the equilibrium of alpha and beta anomers of galactose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALM
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GALM
Gene Accession No. Q66HG4, Q8K157, Q96C23
Gene Alias A530057M15Rik; aldose 1-epimerase; aldose mutarotase; AU015645; AU020959; BLOCK25; epididymis secretory sperm binding protein Li 63p; galactomutarotase; galactose enzyme activator; galactose mutarotase; galactose mutarotase (aldose 1-epimerase); GALM; GLAT; HEL-S-63p; IBD1
Gene Symbols GALM
Host Species Rabbit
Immunogen A synthesized peptide derived from human GALM(Accession Q96C23), corresponding to amino acid residues A264-A314.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130589, 313843, 319625
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.