missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNT12 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97733MFV610
This item is not returnable.
View return policy
Description
GALNT12 Polyclonal antibody specifically detects GALNT12 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GALNT12 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| colorectal cancer, susceptibility to 1, CRCS1, EC 2.4.1.41, FLJ21212, GalNAc-T12, Polypeptide GalNAc transferase 12, polypeptide N-acetylgalactosaminyltransferase 12, pp-GaNTase 12, Protein-UDP acetylgalactosaminyltransferase 12, UDP-GalNAc: polypeptide N-acetylgalactosaminyltransferase, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 12, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 12 (GalNAc-T12) | |
| Produced in rabbits immunized with E. coli-derived Human GALNT12 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 79695 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction