missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNT12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00079695-D02P
This item is not returnable.
View return policy
Description
GALNT12 Polyclonal antibody specifically detects GALNT12 in Human samples. It is validated for Western Blot
Specifications
| GALNT12 | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| AAH13945.1 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| colorectal cancer, susceptibility to 1, CRCS1, EC 2.4.1.41, FLJ21212, GalNAc-T12, Polypeptide GalNAc transferase 12, polypeptide N-acetylgalactosaminyltransferase 12, pp-GaNTase 12, Protein-UDP acetylgalactosaminyltransferase 12, UDP-GalNAc: polypeptide N-acetylgalactosaminyltransferase, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 12, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 12 (GalNAc-T12) | |
| GALNT12 (AAH13945.1, 1 a.a. - 272 a.a.) full-length human protein. MAGGLFAVSKKYFEYLGSYDTGMEVWGGENLEFSFRIWQCGGVLETHPCSHVGHVFPKQAPYSRNKALANSVRAAEVWMDEFKELYYHRNPRARLEPFGDVTERKQLRDKLQCKDFKWFLETVYPELHVPEDRPGFFGMLQNKGLTDYCFDYNPPDENQIVGHQVILYLCHGMGQNQFFEYTSQKEIRYNTHQPEGCIAVEAGMDTLIMHLCEETAPENQKFILQEDGSLFHEQSKKCVQAARKESSDSFVPLLRDCTNSDHQKWFFKERML | |
| 0.1 mg | |
| Stem Cell Markers | |
| 79695 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction