missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNT14 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00079623-B01P
This item is not returnable.
View return policy
Description
GALNT14 Polyclonal antibody specifically detects GALNT14 in Human samples. It is validated for Western Blot
Specifications
| GALNT14 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| EC 2.4.1.41, FLJ12691, FLJ13977, GalNac-T10, GalNac-T14, GALNT15, Polypeptide GalNAc transferase 14, polypeptide N-acetylgalactosaminyltransferase 14, polypeptide N-acetylgalactosaminyltransferase 15, pp-GaNTase 14, Protein-UDP acetylgalactosaminyltransferase 14, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T14, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 14 (GalNAc-T14) | |
| GALNT14 (NP_078848.2, 1 a.a. - 552 a.a.) full-length human protein. MRRLTRRLVLPVFGVLWITVLLFFWVTKRKLEVPTGPEVQTPKPSDADWDDLWDQFDERRYLNAKKWRVGDDPYKLYAFNQRESERISSNRAIPDTRHLRCTLLVYCTDLPPTSIIITFHNEARSTLLRTIRSVLNRTPTHLIREIILVDDFSNDPDDCKQLIKLPKVKCLRNNERQGLVRSRIRGADIAQGTTLTFLDSHCEVNRDWLQPLLHRVKEDYTRVVCPVIDIINLDTFTYIESASELRGGFDWSLHFQWEQLSPEQKARRLDPTEPIRTPIIAGGLFVIDKAWFDYLGKYDMDMDIWGGENFEISFRVWMCGGSLEIVPCSRVGHVFRKKHPYVFPDGNANTYIKNTKRTAEVWMDEYKQYYYAARPFALERPFGNVESRLDLRKNLRCQSFKWYLENIYPELSIPKESSIQKGNIRQRQKCLESQRQNNQETPNLKLSPCAKVKGEDAKSQVWAFTYTQQILQEELCLSVITLFPGAPVVLVLCKNGDDRQQWTKTGSHIEHIASHLCLDTDMFGDGTENGKEIVVNPCESSLMSQHWDMVSS | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NP_078848.2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 79623 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction