missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALNT14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543435
This item is not returnable.
View return policy
Description
Peptide sequence: LEIVPCSRVG HVFRKKHPYV FPDGNANTYI KNTKRTAEVW MDEYKQYYYA Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 79%; Rat: 100%; Zebrafish: 86%.
GALNT14 (EC 2.4.1.41) belongs to a large subfamily of glycosyltransferases residing in the Golgi apparatus. GALNT enzymes catalyze the first step in the O-glycosylation of mammalian proteins by transferring N-acetyl-D-galactosamine (GalNAc) to peptide substrates.
Specifications
| GALNT14 | |
| Polyclonal | |
| Unconjugated | |
| Galnt14 | |
| 0610033M06Rik; FLJ12691; FLJ13977; GalNac-T10; GalNac-T14; GALNT14; GALNT15; Polypeptide GalNAc transferase 14; polypeptide N-acetylgalactosaminyltransferase 14; polypeptide N-acetylgalactosaminyltransferase 15; pp-GaNTase 14; protein-UDP acetylgalactosaminyltransferase 14; si:ch211-147c1.1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 (GalNAc-T14); UNQ2434/PRO4994 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79623 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96FL9 | |
| Galnt14 | |
| Synthetic peptide directed towards the middle region of human GALNT14. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction