missing translation for 'onlineSavingsMsg'
Learn More

GALNT6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15731491 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15731491 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15731491 Supplier Thermo Scientific Supplier No. PA553552

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GALNT6 Polyclonal Antibody

GALNT6 Polyclonal Antibody for Western Blot, ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALNT6
Gene Accession No. Q8NCL4
Gene Alias 4632410F13; AW047994; fc23d02; fc56f12; GalNAc transferase 6; GalNAcT6; galNAc-T6; GALNT6; N-acetylgalactosaminyltransferase T-6; polypeptide GalNAc transferase 6; Polypeptide N-acetylgalactosaminyltransferase 6; pp-GaNTase; pp-GaNTase 6; protein-UDP acetylgalactosaminyltransferase 6; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 (GalNAc-T6); wu:fc23d02; wu:fc56f12; zgc:77836
Gene Symbols GALNT6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALNT6. Recombinant protein control fragment (Product #RP-90275).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11226
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.