missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533575
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Bovine, Horse (94%) Panda, Dog, Bat (89%).
GalR2 expression has been documented widely in the brain and in peripheral tissues including small intestine, heart, kidney, and liver. Localization data from rat CNS indicate high levels of expression in dorsal root ganglia, amygdala, and Purkinje neurons. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Specifications
| GALR2 | |
| Polyclonal | |
| Unconjugated | |
| Galr2 | |
| Gal receptor 2; Gal-2 receptor; GAL2-R; Galanin r2 receptor; galanin receptor 2; galanin receptor type 2; Galanin-2 receptor; Galnr2; GALR2; Gal-r2; GALR-2; mGalR; MGC125983; MGC125984 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O43603 | |
| Galr2 | |
| Synthetic 18 amino acid peptide from 7th transmembrane domain of human GALR2. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction