missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALT Antibody (PSH02-87), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32363
This item is not returnable.
View return policy
Description
GALT Monoclonal antibody specifically detects GALT in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| GALT | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| EC 2.7.7.12, Gal-1-P uridylyltransferase, galactose-1-phosphate uridyl transferase, galactose-1-phosphate uridylyltransferase, UDP-glucose--hexose-1-phosphate uridylyltransferase | |
| Recombinant protein within human GALT aa 1-379/ 379. (Uniprot: P07902) | |
| 100 μL | |
| metabolism | |
| 2592 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| PSH02-87 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction