missing translation for 'onlineSavingsMsg'
Learn More

Galunisertib, MedChemExpress

Product Code. 16476116
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476116

Brand: Medchem Express HY13226

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Galunisertib (LY2157299) is an oral and selective TGF-β receptor type I (TGF-βRI) kinase inhibitor with an IC50 of 56 nM.

Galunisertib (LY2157299) is an oral and selective TGF-β receptor type I (TGF-βRI) kinase inhibitor with an IC50 of 56 nM.

Specifications

Chemical Name or Material Galunisertib
CAS 700874-72-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H19N5O
Quantity 100 mg
Synonym LY2157299
Solubility Information DMSO : 100 mg/mL (270.69 mM; Need ultrasonic)
SMILES CC1=CC=CC(C2=NN3C(CCC3)=C2C4=CC=NC5=C4C=C(C=C5)C(N)=O)=N1
Molecular Weight (g/mol) 369.42
Formula Weight 369.42
Percent Purity 98.65%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.