Learn More
Galvinoxyl Free Radical, TRC
Specifications
Specifications
| Chemical Name or Material | Galvinoxyl Free Radical |
| CAS | 2370-18-5 |
| Molecular Formula | C29H41O2 |
| Quantity | 250 mg |
| Synonym | Galvinoxy, Galvinoxyl, Galvinoxyl Radical, Coppinger's Radical, 4-[[3,5-Bis(1,1-dimethylethyl)-4-oxo-2,5-cyclohexadien-1-ylidene]methyl]-2,6-bis(1,1-dimethylethyl)phenoxy, 2,6-Di-tert-butyl-alpha-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy, 2,6-di-tert-butyl-alpha-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy, 4-[[3,5-bis(1,1-dimethylethyl)-4-oxo-2,5-cyclohexadien-1-ylidene]methyl]-2,6-bis(1,1-dimethylethyl)phenoxy |
| InChI Formula | InChI=1S/C29H41O2/c1-18(2)9-24-14-22(15-25(28(24)30)10-19(3)4)13-23-16-26(11-20(5)6)29(31)27(17-23)12-21(7)8/h13-21H,9-12H2,1-8H3 |
| SMILES | CC(C)CC1=CC(=Cc2cc(CC(C)C)c([O])c(CC(C)C)c2)C=C(CC(C)C)C1=O |
| IUPAC Name | 4-[[4-λ^{1}-oxidanyl-3,5-bis(2-methylpropyl)phenyl]methylidene]-2,6-bis(2-methylpropyl)cyclohexa-2,5-dien-1-one |
| Molecular Weight (g/mol) | 421.63 |
| Formula Weight | 421.3107 |