missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gamithromycin, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Description
Gamithromycin is an antimicrobial agent which can inhibit the growth of MmmSC strains B237 and Tan8 with MICs of 0.00012 and 0.00006 μg/mL, respectively.
Specifications
Specifications
| Chemical Name or Material | Gamithromycin |
| Physical Form | Solid |
| CAS | 145435-72-9 |
| Color | White |
| Quantity | 2 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C40H76N2O12 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | ML-1709460 |
| Solubility Information | DMSO : 100 mg/mL (128.69 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction