Learn More
gamma-1,2,3,4,5,6-Hexachlorocyclohexane-d6, CDN
Product Code. 30323011
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323011
Brand: CDN CDND30100.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material gamma-HCH D6
CAS 60556-82-3
Molecular Formula C6 2H6 Cl6
Quantity 0.005 g
Synonym (1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane-1,2,3,4,5,6-d6, Hexadeuterated lindane, Lindane-d6, gamma-HCH D6
InChI Formula InChI=1 S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6 H/t1-,2-,3-,4+,5+,6+/i1D,2 D,3 D,4 D,5 D,6 D
SMILES [2 H][C@@]1(Cl)[C@@]([2 H])(Cl)[C@@]([2 H])(Cl)[C@]([2 H])(Cl)[C@@]([2 H])(Cl)[C@@]1([2 H])Cl
IUPAC Name 1,2,3,4,5,6-hexachloro-1,2,3,4,5,6-hexadeuteriocyclohexane
Molecular Weight (g/mol) 296.87
Formula Weight 293.8977 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity