missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Catenin Antibody (CTNG/1664), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54524PE
This item is not returnable.
View return policy
Description
gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| gamma Catenin | |
| Monoclonal | |
| PE | |
| catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
| Recombinant full-length human gamma Catenin protein (Uniprot: P14923) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 3728 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CTNG/1664 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction