missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ gamma Catenin Polyclonal Antibody, Biotin

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142008
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise both reported isoforms, as represented by NP_002221.1 and NP_068831.1. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.
Specifications
| gamma Catenin | |
| Polyclonal | |
| Biotin | |
| JUP | |
| ARVD12; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; cb80; cell-adhesion protein plakoglobin; Ctnng; D930025P04Rik; danPG; Desmoplakin III; desmoplakin-3; desmosomal protein; DP3; DPIII; fb11b06; gama-catenin; gamma-catenin; gamma-catenin (plakoglobin); junction plakoglobin; junction plakoglobin a; Jup; jup protein; jupa; OTTHUMP00000164735; PDGB; PG; PKGB; plakoglobin; wu:fb11b06 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 16480, 3728, 81679 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P14923, Q02257, Q6P0K8 | |
| JUP | |
| Peptide with sequence C-GLRPPYPTADHMLA (aa 732-745). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction