Learn More
Keystone Cleanroom Products Gamma-Irradiated Heat-Seal Sterility Maintenance Bag
Bag used to cover wrapped items after sterilization to provide protection from environmental factors that could compromise sterility.
Brand: Keystone Cleanroom Products 3234ST
Additional Details : Weight : 17.21380kg
Description
- Made of a transparent, strong, 3 mil thick medical grade polyolefin material
- Features a starter slit to facilitate easy opening of the cover without the need for cutting instruments which could damage the contents
- Gamma irradiated
- Lot controlled
- Ready to use
- Packaged in easy to open Tyvek™ peel pouches
- Minimize contamination risks
- Simplifies conformance to SOP's
- Eliminate the need for wiping down contaminated writing instruments
Specifications
Not Autoclavable | |
COC, Certificate of Irradiation | |
Clear | |
76.2 μm | |
76.2 x 61 cm (24 x 30 in.) |
20.4 L | |
Heat Seal | |
Polyolefin | |
3 mil |