missing translation for 'onlineSavingsMsg'
Learn More

gamma-secretase modulator 3, MedChemExpress

Product Code. 16477886
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477886 100 mg 100mg
16418138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477886 Supplier Medchem Express Supplier No. HY50889

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress gamma-secretase modulator 3 is a gamma-secretase modulator.

gamma-secretase modulator 3 is a gamma-secretase modulator.

Specifications

Chemical Name or Material gamma-secretase modulator 3
CAS 1431697-84-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H23FN4OS
Quantity 100 mg
Solubility Information DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES COC1=C(N2C=C(C)N=C2)C=CC(NC3=NC4=C(CCCC4C5=CC=C(F)C=C5)S3)=C1
Molecular Weight (g/mol) 434.53
Formula Weight 434.53
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.