Learn More
Gamma-Tocopherol-d4 (major), TRC
Specifications
Specifications
| Chemical Name or Material | gamma-Tocopherol-d4 |
| CAS | 1329652-13-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28H44D4O2 |
| Quantity | 10 mg |
| Synonym | (2R)-3,4-Dihydro-2,8-dimethyl-7-(methyl-d4)-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol, (+)- 2,8-Dimethyl-(7-methyl-d4)-2-(4,8,12-trimethyltridecyl) -6-chromanol, (+)-γ-Tocopherol-d4, (2R,4'R,8'R)-γ-Tocopherol-d4, (R,R,R)-γ -Tocopherol-d4, D-γ-Tocopherol-d4, all-(R)-γ-Tocopherol-d4, d-γ-Tocopherol-d4 |
| InChI Formula | InChI=1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-19-26(29)23(5)24(6)27(25)30-28/h19-22,29H,8-18H2,1-7H3/t21-,22-,28-/m1/s1/i5D3,19D |
| SMILES | OC1=C([2H])C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O2)=C2C(C)=C1C([2H])([2H])[2H] |
| Molecular Weight (g/mol) | 420.7 |
| Formula Weight | 420.3905 |
| Show More |