missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Antibody (9D6), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60245AF532
This item is not returnable.
View return policy
Description
gamma Tubulin Monoclonal antibody specifically detects gamma Tubulin in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| gamma Tubulin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| Recombinant human gamma Tubulin (1-451aa) purified from E. coli. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Centrosome Markers | |
| 7283 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunofluorescence | |
| 9D6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction