missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99067C
This item is not returnable.
View return policy
Description
gamma Tubulin Polyclonal antibody specifically detects gamma Tubulin in Human samples. It is validated for Western Blot
Specifications
| gamma Tubulin | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human gamma Tubulin. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Centrosome Markers | |
| 7283 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction