Learn More
gamma Tubulin Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99067F
Product Code. 18655485
  • € 407.00 / 0.10mL
Description
gamma Tubulin Polyclonal antibody specifically detects gamma Tubulin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
gamma Tubulin
Polyclonal
PBS
Store at 4C in the dark.