missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Antibody (TU-32), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-90616H
This item is not returnable.
View return policy
Description
gamma Tubulin Monoclonal antibody specifically detects gamma Tubulin in Human, Porcine, Animal, Plant, Protozoa samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| gamma Tubulin | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Pig, Animal species, Plant, Protozoa | |
| Purified |
| Western Blot, Immunocytochemistry | |
| TU-32 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| human gamma-tubulin peptide EYHAATRPDYISWGTQ, amino acids 434-449 | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Centrosome Markers | |
| 7283 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction