missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ gamma Tubulin Monoclonal Antibody (10-A4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544601
This item is not returnable.
View return policy
Description
gamma Tubulin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
Specifications
| gamma Tubulin | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P23258, P83887, P83888, Q8VCK3, Q9NRH3 | |
| TUBG1, Tubg2 | |
| Synthetic peptide within Human gamma tubulin aa 402-451/451. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 10-A4 | |
| Unconjugated | |
| TUBG1 | |
| 1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; centrosome marker; gamma tubulin; gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; Tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; Unknown (protein for MGC:133882); XELAEV_18002398mg; xgam; zgc:55894 | |
| Mouse | |
| Protein G | |
| RUO | |
| 103733, 103768, 252921, 27175, 680991, 7283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction