Learn More
Ansell GAMMEX™ Latex Textured Surgical Gloves
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Description
- ASTM D3577 certified
- Micro-textured finish with special external surface treatment to ensure exceptional grip under dry and damp conditions
- Ideal thickness to optimize tactile sensitivity and protection
- DERMASHIELD™ Technology internal polymer coating facilitates dry and damp donning as well as intra-operative changes
Specifications
Certifications/Compliance | ASTM D3577, ISO 10282 |
Certifications/Compliance (Europe) | EN 16523-1, EN 374-2 and -4, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, EN 420:2003 + A1:2009 |
Accelerator Free | No |
Applications | Healthcare |
AQL | 0.65 |
Color | Natural |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Finger Thickness (English) | 8.66 ±0.79 mil |
Finger Thickness (Metric) | 0.22 ±0.02 mm |