Learn More
GAMT Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-34854G
Product Code. 18790951
  • € 633.00 / 0.10mL
Description
GAMT Polyclonal antibody specifically detects GAMT in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Guanidinoacetate N-methyltransferase.
RUO
Primary
Western Blot
Immunogen affinity purified
Proteases & Other Enzymes