missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAMT Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34854JF646
This item is not returnable.
View return policy
Description
GAMT Polyclonal antibody specifically detects GAMT in Human samples. It is validated for Western Blot
Specifications
| GAMT | |
| Polyclonal | |
| Western Blot | |
| EC 2.1.1.2, guanidinoacetate N-methyltransferase, PIG2, TP53I2 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Guanidinoacetate N-methyltransferase. | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 2593 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction