missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GANAB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584496
This item is not returnable.
View return policy
Description
Immunogen sequence: GRDENSVELT MAEGPYKIIL TARPFRLDLL EDRSLLLSVN ARGLLEFEHQ RAPRVSQGSK DPAEGDGAQP EETPRDG Highest antigen sequence indentity to the following orthologs: Mouse - 68%, Rat - 35%.
This gene encodes the alpha subunit of glucosidase II and a member of the glycosyl hydrolase 31 family of proteins. The heterodimeric enzyme glucosidase II plays a role in protein folding and quality control by cleaving glucose residues from immature glycoproteins in the endoplasmic reticulum. Expression of the encoded protein is elevated in lung tumor tissue and in response to UV irradiation. Mutations in this gene cause autosomal-dominant polycystic kidney and liver disease.
Specifications
| GANAB | |
| Polyclonal | |
| Unconjugated | |
| Ganab | |
| alpha glucosidase 2 alpha neutral subunit; alpha-glucosidase 2; AU042638; carbohydrate processing enzyme of the endoplasmic reticulum; G2an; GANAB; GIIA; glucosidase II alpha subunit; glucosidase II alpha-subunit; glucosidase II subunit alpha; glucosidase, alpha; neutral AB; GLUII; KIAA0088; mKIAA0088; neutral AB; neutral alpha-glucosidase AB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23193 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14697 | |
| Ganab | |
| Recombinant protein corresponding to Human GANAB. Recombinant protein control fragment (Product #RP-104098). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction