missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GANC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 433.00 - € 572.00
Specifications
| Antigen | GANC |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18411781
|
Novus Biologicals
NBP1-86448-25ul |
25 μL |
€ 433.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18265997
|
Novus Biologicals
NBP1-86448 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GANC Polyclonal specifically detects GANC in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GANC | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 3.2.1, EC 3.2.1.20, glucosidase, alpha; neutral C, MGC138256, neutral alpha-glucosidase C | |
| GANC | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2595 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ASQPVMRPLWVEFPDELKTFDMEDEYMLGSALLVHPVTEPKATTVDVFLPGSNEVWYDYKTFAHWEGGCTVKIPVALDTIPVFQRGGSVIPIKTTVGKSTGWMTESSYGLRVALSTKGSSVGELYLDDGHSFQYLHQKQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title