missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GANC Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582567
This item is not returnable.
View return policy
Description
Immunogen sequence: ASQPVMRPLW VEFPDELKTF DMEDEYMLGS ALLVHPVTEP KATTVDVFLP GSNEVWYDYK TFAHWEGGCT VKIPVALDTI PVFQRGGSVI PIKTTVGKST GWMTESSYGL RVALSTKGSS VGELYLDDGH SFQYLHQKQ Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 81%.
Glycosyl hydrolase enzymes hydrolyse the glycosidic bond between two or more carbohydrates, or between a carbohydrate and a non-carbohydrate moiety. This gene encodes a member of glycosyl hydrolases family 31. This enzyme hydrolyses terminal, non-reducing 1,4-linked alpha-D-glucose residues and releases alpha-D-glucose. This is a key enzyme in glycogen metabolism and its gene localizes to a chromosomal region (15q15) that is associated with susceptibility to diabetes.
Specifications
| GANC | |
| Polyclonal | |
| Unconjugated | |
| GANC | |
| 5830445O15Rik; 9330160A12; Ganc; glucosidase alpha, neutral C; glucosidase, alpha; neutral C; mFLJ00088; neutral alpha-glucosidase C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2595 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TET4 | |
| GANC | |
| Recombinant protein corresponding to Human GANC. Recombinant protein control fragment (Product #RP-89238). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction