missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GANC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598675
This item is not returnable.
View return policy
Description
GANC Polyclonal Antibody for Western Blot, IHC (P), ELISA
Glycosyl hydrolase enzymes hydrolyse the glycosidic bond between two or more carbohydrates, or between a carbohydrate and a non-carbohydrate moiety. This gene encodes a member of glycosyl hydrolases family 31. This enzyme hydrolyses terminal, non-reducing 1,4-linked alpha-D-glucose residues and releases alpha-D-glucose. This is a key enzyme in glycogen metabolism and its gene localizes to a chromosomal region (15q15) that is associated with susceptibility to diabetes.
Specifications
| GANC | |
| Polyclonal | |
| Unconjugated | |
| GANC | |
| 5830445O15Rik; 9330160A12; Ganc; glucosidase alpha, neutral C; glucosidase, alpha; neutral C; mFLJ00088; neutral alpha-glucosidase C | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2595 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4.29 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8TET4 | |
| GANC | |
| Recombinant Human Neutral alpha-glucosidase C protein (171-318AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction