missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gankyrin Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1834B
This item is not returnable.
View return policy
Description
Gankyrin Polyclonal specifically detects Gankyrin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifications
| Gankyrin | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular) | |
| O75832 | |
| PSMD10 | |
| A synthetic peptide made to an internal portion of the human Gankyrin protein sequence (between residues 100-200). [UniProt# O75832] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted to react with mouse, rat, porcine, bovine, and zebrafish based on 100% sequence homology. | |
| Store at 4C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 | |
| Rabbit | |
| 24 kDa | |
| 0.1 mL | |
| Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| 5716 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction