missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gankyrin Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1834UV
This item is not returnable.
View return policy
Description
Gankyrin Polyclonal specifically detects Gankyrin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifications
| Gankyrin | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular) | |
| 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 | |
| Rabbit | |
| 0.1 mL | |
| 5716 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PSMD10 | |
| A synthetic peptide made to an internal portion of the human Gankyrin protein sequence (between residues 100-200). [UniProtNo. O75832] | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction