missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gankyrin Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1834FR
This item is not returnable.
View return policy
Description
Gankyrin Polyclonal antibody specifically detects Gankyrin in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular)
Specifications
| Gankyrin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| DyLight 680 | |
| 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 | |
| A synthetic peptide made to an internal portion of the human Gankyrin protein sequence (between residues 100-200). [UniProt# O75832] | |
| 0.1 mL | |
| Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| 5716 | |
| Store at 4°C in the dark. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur