missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GANP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583220
This item is not returnable.
View return policy
Description
Immunogen sequence: RDWYDFVWNR TRGIRKDITQ QHLCDPLTVS LIEKCTRFHI HCAHFMCEEP MSSFDAKINN ENMTKCLQSL KEMYQDLRNK GVFCASE Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
POLE3 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.
Specifications
| GANP | |
| Polyclonal | |
| Unconjugated | |
| MCM3AP | |
| 80 kDa MCM3-associated protein; FLJ44336; FLJ45306; GANP; GC-associated DNA primase; germinal center-associated nuclear protein; germinal-center associated nuclear protein; germinal-centre associated nuclear protein; KIAA0572; MAP80; MCM3 acetylating protein; MCM3 acetyltransferase; MCM3 import protein; MCM3 minichromosome maintenance deficient 3 associated protein; MCM3AP; minichromosome maintenance complex component 3 associated protein; minichromosome maintenance deficient 3 (S. cerevisiae) associated protein; minichromosome maintenance deficient 3 associated protein; minichromosome maintenance deficient 3-associated protein; mKIAA0572; nuclear protein GANP; SAC3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8888 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60318 | |
| MCM3AP | |
| Recombinant protein corresponding to Human MCM3AP. Recombinant protein control fragment (Product #RP-105881). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction