missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies GANT61
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
- GANT61 is a hexahydropyrimidine derivative which selectively inhibits GLI transcription factors in the Hedgehog signaling pathway
- GANT61 acts downstream of Smoothened to inhibit both GLI1- and GLI2-mediated transcription (IC50 = 5 μM in a GLI-transfected cell line) by preventing their DNA binding (Lauth et al.)
- Inhibits in vitro proliferation of cancer cell lines (PANC1 and 22Rv1) with elevated GLI1 levels, and prevents the development of 22Rv1 tumors in mice (Lauth et al.)
- Inhibits pancreatic cancer stem cell growth in vitro and in mouse xenograft model (Fu et al.)
- Induces cell death of primitive neuroectodermal tumor cell line in a caspase-independent manner by inhibiting DNA replication (Matsumoto et al.)
- Causes apoptosis in myeloid leukemia cells and in some cell lines is more effective when combined with rapamycin (Pan et al.)
Specifications
Specifications
| CAS | 500579-04-4 |
| Synonym | NSC 136476 |
| Quantity | 10 mg |
| Molecular Formula | C27H35N5 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction