Learn More
Invitrogen™ Gantenerumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559077
 View more versions of this product
Product Code. 30283244
  • € 477.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Gantenerumab is under investigation for the treatment of Dementia, Alzheimer's Disease, and Alzheimer's Disease, Familial.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gantenerumab
Recombinant Monoclonal
Unconjugated
R1450
Protein A
RUO
Human
Antibody
IgG1 κ
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized