Learn More
Invitrogen™ GAP43 Monoclonal Antibody (1E3)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA547386
 View more versions of this product
Product Code. 17895117
  • € 504.00 / 100µL
Description
GAP43 Monoclonal Antibody for Western Blot, ICC/IF, IHC
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile growth cones that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GAP43
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
P06837, P07936, P17677
GAP43
C-terminal peptide of rat and mouse GAP43, which is KEDPEADQEHA, with an N-terminal Cys added to allow chemical coupling to KLH carrier protein.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry, Western Blot, Immunocytochemistry
1E3
Unconjugated
GAP43
Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1
Mouse
Protein A/G
RUO
14432, 2596, 29423
-20°C, Avoid Freeze/Thaw Cycles