missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAP43 Monoclonal Antibody (5E8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ PA5143534
This item is not returnable.
View return policy
Description
GAP43 Monoclonal Antibody for Western Blot, ICC/IF, IHC
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile growth cones that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
| GAP43 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P06837, P07936, P17677 | |
| GAP43 | |
| Recombinant full-length Human GAP43 expressed in and purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 5E8 | |
| Unconjugated | |
| GAP43 | |
| Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 14432, 2596, 29423 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction