missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAP43 Polyclonal Antibody, HRP

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522898
This item is not returnable.
View return policy
Description
This antibody does not react with xenopus samples. Suggested positive control: embryonic nervous tissue, brain, injured nerve. Immunogen sequence: KEDPEADQEH A.
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
| GAP43 | |
| Polyclonal | |
| HRP | |
| GAP43 | |
| Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100061125, 100623423, 14432, 2596, 281777, 29423, 427955, 478572 | |
| 4°C, store in dark | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.79 mg/mL | |
| PBS with no preservative | |
| P06836, P06837, P07936, P17677, P35001 | |
| GAP43 | |
| C-terminal peptide of rodent GAP43 coupled to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Pig, Horse, Chicken, Drosophila | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction