missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAP43 Polyclonal Antibody

Chicken Polyclonal Antibody
Brand: Invitrogen™ PA595660
This item is not returnable.
View return policy
Description
GAP43 Polyclonal Antibody for Western Blot, ICC/IF, IHC
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
| GAP43 | |
| Polyclonal | |
| Unconjugated | |
| GAP43 | |
| Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
| Chicken | |
| IgY fraction | |
| RUO | |
| 100061125, 14432, 2596, 281777, 29423 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 20 mg/mL | |
| PBS with 10mM sodium azide | |
| P06836, P06837, P07936, P17677 | |
| GAP43 | |
| Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Horse | |
| Antibody | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction