missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH Antibody (GA1R), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-37828CL1
This item is not returnable.
View return policy
Description
GAPDH Monoclonal antibody specifically detects GAPDH in Bacteria,Chicken,Hamster,Human,Insect,Mouse,Porcine,Rabbit,Rat,Yeast samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Simple Western,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| GAPDH | |
| Monoclonal | |
| CoraFluor 1 | |
| aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
| This GAPDH protein was developed against recombinant GAPDH | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Autophagy, Cancer, Chromatin Research, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
| 2597 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| GA1R | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bacteria, Chicken, Hamster, Human, Insect | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction