Learn More
Garcin™ Garcin™ Bactinyl Wipers
Click to view available options
Description:
Odorless
Scented
Unit Size:
Pack of 12
Description
- Fragranced or odorless
- Rechargeable dispenser box with reclosable case
- Active substances include peroxides, quaternary ammoniums, ethanol
- Mineral-certified impregnation solution
Specifications
Specifications
| Product Type | Wipers |
| Length (Metric) | 13 cm |
| Width (Metric) | 20 cm |
| Description | Scented |
| Packaging Quantity | 12 Pk. |